Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKdZeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128652.4 | NP_180655.1 | 9 - 10 | 1058 | 98 | |
GenBank mRNA | AY064020.1 | - | 9 - 10 | 1008 | 98 | |
AY094423.1 | - | 9 - 10 | 1034 | 98 | ||
AY096698.1 | - | 9 - 10 | 955 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |