Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPUORF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036376.1 | NP_001031453.1 | 2 - 3 | 173 | 69 | |
NM_001036377.1 | NP_001031454.1 | 1 - 2 | 686 | 69 | ||
NM_128684.5 | NP_180686.2 | 1 - 2 | 955 | 69 | ||
GenBank mRNA | AJ576042.1 | - | 2 - 3 | 173 | 69 | |
AY954808.1 | - | 1 - 2 | 95 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |