Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336351.1 | NP_001318329.1 | 18 - 19 | 2671 | 89 | |
NM_001336352.1 | NP_001323600.1 | 16 - 17 | 1958 | 89 | ||
NM_001336353.1 | NP_001323601.1 | 11 - 12 | 1184 | 89 | ||
NM_001336354.1 | NP_001323599.1 | 14 - 15 | 1749 | 89 | ||
NM_128748.3 | NP_180749.2 | 16 - 17 | 1958 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |