Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLB01 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128820.3 | NP_180820.2 | 1 - 2 | 384 | 83 | |
GenBank mRNA | KJ138879.1 | - | 1 - 2 | 267 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |