Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336434.1 | NP_001324212.1 | 40 - 41 | 5540 | 133 | |
NM_001336435.1 | NP_001324208.1 | 40 - 41 | 5086 | 133 | ||
NM_001336436.1 | NP_001324211.1 | 40 - 41 | 5107 | 133 | ||
NM_001336437.1 | NP_001324213.1 | 26 - 27 | 3597 | 133 | ||
NM_001336438.1 | NP_001324210.1 | 40 - 41 | 5368 | 133 | ||
NM_001336439.1 | NP_001324209.1 | 41 - 42 | 5062 | 133 | ||
NM_001336440.1 | NP_001324214.1 | 41 - 42 | 5062 | 133 | ||
NM_128883.3 | NP_180882.2 | 40 - 41 | 5125 | 133 | ||
GenBank mRNA | LC177513.1 | - | 39 - 40 | 5053 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |