Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACHT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128887.3 | NP_180885.1 | 2 - 3 | 571 | 136 | |
GenBank mRNA | DQ056563.1 | - | 2 - 3 | 405 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |