Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128949.3 | NP_565776.1 | 9 - 10 | 1436 | 65 | |
GenBank mRNA | AF360290.1 | - | 9 - 10 | 1358 | 65 | |
AK229595.1 | - | 8 - 9 | 1231 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Arabidopsis thaliana Probe spans exons |