Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198270.1 | NP_001185199.1 | 3 - 4 | 437 | 82 | |
NM_001333494.1 | NP_001322512.1 | 1 - 2 | 2333 | 82 | ||
NM_104047.4 | NP_564595.1 | 2 - 3 | 2062 | 82 | ||
NM_202274.2 | NP_974003.1 | 1 - 2 | 359 | 82 | ||
GenBank mRNA | AK230310.1 | - | 2 - 3 | 1842 | 82 | |
AY120756.1 | - | 1 - 2 | 357 | 82 | ||
BT000109.1 | - | 1 - 2 | 115 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |