Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084229.1 | NP_001077698.1 | 12 - 13 | 1083 | 96 | |
NM_104049.4 | NP_564596.1 | 14 - 15 | 1497 | 96 | ||
GenBank mRNA | AY050817.1 | - | 14 - 15 | 1261 | 96 | |
AY084730.1 | - | 14 - 15 | 1265 | 96 | ||
AY114081.1 | - | 13 - 14 | 1196 | 96 | ||
BT000658.1 | - | 14 - 15 | 1266 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |