Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116156.5 | NP_191850.2 | 11 - 12 | 1087 | 94 | |
GenBank mRNA | AB109893.1 | - | 11 - 12 | 993 | 94 | |
AY090253.1 | - | 11 - 12 | 1034 | 94 | ||
AY125535.1 | - | 11 - 12 | 993 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |