Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084865.2 | NP_001078334.1 | 10 - 11 | 1743 | 102 | |
NM_116183.4 | NP_191877.2 | 10 - 11 | 1790 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |