Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036498.3 | NP_001031575.1 | 7 - 8 | 905 | 84 | |
NM_001036499.1 | NP_001031576.1 | 7 - 8 | 841 | 84 | ||
NM_001203732.1 | NP_001190661.1 | 7 - 8 | 903 | 84 | ||
NM_116491.3 | NP_567243.1 | 6 - 7 | 999 | 84 | ||
GenBank mRNA | AJ318017.1 | - | 5 - 6 | 700 | 84 | |
AK317123.1 | - | 7 - 8 | 842 | 84 | ||
AY046030.1 | - | 6 - 7 | 922 | 84 | ||
AY133878.1 | - | 5 - 6 | 666 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |