Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203746.1 | NP_001190675.1 | 10 - 11 | 1597 | 158 | |
NM_116723.5 | NP_567272.4 | 10 - 11 | 1378 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |