Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340521.1 | NP_001328262.1 | 8 - 9 | 723 | 132 | |
NM_116734.3 | NP_567276.1 | 9 - 10 | 880 | 132 | ||
GenBank mRNA | AY045901.1 | - | 9 - 10 | 808 | 132 | |
AY113948.1 | - | 9 - 10 | 787 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |