Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116736.2 | NP_567278.1 | 1 - 2 | 1126 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |