Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR3;5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121965.4 | NP_568377.1 | 6 - 7 | 1163 | 77 | |
GenBank mRNA | AB061739.1 | - | 6 - 7 | 1113 | 77 | |
AK226785.1 | - | 6 - 7 | 1121 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |