Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABG3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104107.3 | NP_175638.1 | 5 - 6 | 539 | 118 | |
GenBank mRNA | AY074361.1 | - | 5 - 6 | 539 | 118 | |
AY096407.1 | - | 5 - 6 | 402 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |