Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMH-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333547.1 | NP_001322993.1 | 3 - 4 | 500 | 97 | |
NM_001333548.1 | NP_001322994.1 | 3 - 4 | 459 | 97 | ||
NM_001333549.1 | NP_001322995.1 | 3 - 4 | 459 | 97 | ||
NM_104128.3 | NP_564608.1 | 3 - 4 | 500 | 97 | ||
NM_179467.3 | NP_849798.1 | 3 - 4 | 412 | 97 | ||
GenBank mRNA | AF099971.1 | - | 3 - 4 | 387 | 97 | |
BT043496.1 | - | 3 - 4 | 337 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |