Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104225.2 | NP_175752.1 | 3 - 4 | 2346 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |