Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333607.1 | NP_001321367.1 | 2 - 3 | 217 | 86 | |
NM_104250.5 | NP_175776.2 | 2 - 3 | 224 | 86 | ||
GenBank mRNA | AY568526.1 | - | 1 - 2 | 98 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |