Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333641.1 | NP_001321996.1 | 13 - 14 | 1384 | 83 | |
NM_104312.3 | NP_175836.1 | 14 - 15 | 1346 | 83 | ||
GenBank mRNA | AF316501.1 | - | 14 - 15 | 1172 | 83 | |
BT025983.1 | - | 14 - 15 | 1152 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |