Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333641.1 | NP_001321996.1 | 7 - 8 | 604 | 129 | |
NM_104312.3 | NP_175836.1 | 7 - 8 | 671 | 129 | ||
GenBank mRNA | AF316501.1 | - | 7 - 8 | 497 | 129 | |
BT025983.1 | - | 7 - 8 | 477 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |