Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198295.1 | NP_001185224.1 | 13 - 14 | 859 | 76 | |
NM_001198296.1 | NP_001185225.1 | 13 - 14 | 794 | 76 | ||
NM_001333653.1 | NP_001319226.1 | 12 - 13 | 1202 | 76 | ||
NM_001333654.1 | NP_001322059.1 | 3 - 4 | 720 | 76 | ||
NM_104329.5 | NP_175853.1 | 13 - 14 | 955 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |