Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333684.1 | NP_001319236.1 | 12 - 13 | 1229 | 86 | |
NM_104370.5 | NP_175894.5 | 12 - 13 | 1205 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |