Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333873.1 | NP_001320771.1 | 8 - 9 | 1448 | 79 | |
NM_104690.4 | NP_176206.2 | 9 - 10 | 1399 | 79 | ||
GenBank mRNA | AK118901.1 | - | 9 - 10 | 1295 | 79 | |
BT005502.1 | - | 9 - 10 | 1019 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |