Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333894.1 | NP_001320517.1 | 8 - 9 | 1217 | 82 | |
NM_001333895.1 | NP_001320518.1 | 8 - 9 | 981 | 82 | ||
NM_104731.5 | NP_176247.2 | 9 - 10 | 1113 | 82 | ||
GenBank mRNA | AK229327.1 | - | 9 - 10 | 1096 | 82 | |
BT011005.1 | - | 9 - 10 | 963 | 82 | ||
BT015384.1 | - | 9 - 10 | 925 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |