Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GolS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104732.2 | NP_176248.1 | 1 - 2 | 476 | 122 | |
GenBank mRNA | DQ056502.1 | - | 1 - 2 | 323 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |