Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPAC43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104745.3 | NP_176261.1 | 1 - 2 | 492 | 66 | |
GenBank mRNA | AF386969.1 | - | 1 - 2 | 249 | 66 | |
BT001161.1 | - | 1 - 2 | 161 | 66 | ||
L34773.1 | - | 1 - 2 | 280 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |