Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS04 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104793.3 | NP_564772.1 | 6 - 7 | 1306 | 69 | |
GenBank mRNA | AY059757.1 | - | 6 - 7 | 1304 | 69 | |
BT001960.1 | - | 6 - 7 | 1252 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |