Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104910.3 | NP_564792.1 | 3 - 4 | 956 | 68 | |
GenBank mRNA | AB493514.1 | - | 3 - 4 | 617 | 68 | |
AF331712.1 | - | 3 - 4 | 673 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |