Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336523.1 | NP_001324697.1 | 33 - 34 | 5146 | 74 | |
NM_129022.8 | NP_181015.7 | 33 - 34 | 4957 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |