Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129025.4 | NP_181018.1 | 11 - 12 | 1973 | 65 | |
GenBank mRNA | AJ441291.1 | - | 11 - 12 | 1611 | 65 | |
AY099791.1 | - | 11 - 12 | 1889 | 65 | ||
BT000335.1 | - | 11 - 12 | 1611 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |