Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129032.5 | NP_181025.1 | 1 - 2 | 651 | 80 | |
GenBank mRNA | BT004022.1 | - | 1 - 2 | 523 | 80 | |
BT005187.1 | - | 1 - 2 | 490 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |