Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129043.5 | NP_181036.2 | 5 - 6 | 769 | 154 | |
NM_201873.2 | NP_973602.1 | 5 - 6 | 716 | 154 | ||
GenBank mRNA | AK230434.1 | - | 5 - 6 | 731 | 154 | |
AY180100.1 | - | 5 - 6 | 647 | 154 | ||
BT026469.1 | - | 5 - 6 | 647 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |