Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084537.1 | NP_001078006.1 | 1 - 2 | 370 | 133 | |
NM_129053.6 | NP_181046.1 | 1 - 2 | 407 | 133 | ||
GenBank mRNA | BT025773.1 | - | 1 - 2 | 243 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |