Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336552.1 | NP_001323587.1 | 11 - 12 | 726 | 124 | |
NM_129086.4 | NP_181077.3 | 15 - 16 | 1286 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |