Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198373.2 | NP_001185302.1 | 3 - 4 | 635 | 101 | |
NM_105042.5 | NP_176552.1 | 2 - 3 | 722 | 101 | ||
NM_202351.2 | NP_974080.1 | 3 - 4 | 470 | 101 | ||
GenBank mRNA | AB493517.1 | - | 2 - 3 | 392 | 101 | |
AF251687.1 | - | 2 - 3 | 392 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |