Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPO11-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334143.1 | NP_001320320.1 | 5 - 6 | 559 | 65 | |
NM_105072.3 | NP_176582.2 | 5 - 6 | 559 | 65 | ||
GenBank mRNA | AJ251990.1 | - | 5 - 6 | 548 | 65 | |
DQ446392.1 | - | 5 - 6 | 525 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |