Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105119.5 | NP_176625.1 | 7 - 8 | 991 | 68 | |
GenBank mRNA | BT011226.1 | - | 7 - 8 | 903 | 68 | |
BT012154.1 | - | 7 - 8 | 864 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |