Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035536.3 | NP_001030613.1 | 8 - 9 | 1763 | 79 | |
NM_001337314.1 | NP_001327194.1 | 7 - 8 | 1761 | 79 | ||
NM_001337315.1 | NP_001327195.1 | 8 - 9 | 1837 | 79 | ||
NM_110975.5 | NP_186759.2 | 8 - 9 | 1763 | 79 | ||
GenBank mRNA | BT002498.1 | - | 8 - 9 | 1632 | 79 | |
BT008419.1 | - | 7 - 8 | 1477 | 79 | ||
KJ920366.1 | - | 7 - 8 | 1477 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |