Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340813.1 | NP_001329616.1 | 5 - 6 | 920 | 89 | |
NM_117360.8 | NP_193027.5 | 5 - 6 | 963 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |