Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117479.4 | NP_193140.1 | 1 - 2 | 410 | 68 | |
GenBank mRNA | AF370130.1 | - | 1 - 2 | 185 | 68 | |
AY040046.1 | - | 1 - 2 | 144 | 68 | ||
AY065104.1 | - | 1 - 2 | 179 | 68 | ||
AY065108.1 | - | 1 - 2 | 187 | 68 | ||
BT000385.1 | - | 1 - 2 | 144 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |