Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLB06 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341028.1 | NP_001328692.1 | 11 - 12 | 2160 | 155 | |
NM_117620.1 | NP_193267.1 | 11 - 12 | 2289 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Arabidopsis thaliana Probe spans exons |