Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLB06 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341028.1 | NP_001328692.1 | 5 - 6 | 1175 | 135 | |
NM_001341029.1 | NP_001328691.1 | 5 - 6 | 1081 | 135 | ||
NM_001341030.1 | NP_001328693.1 | 5 - 6 | 1107 | 135 | ||
NM_001341031.1 | NP_001328694.1 | 5 - 6 | 1037 | 135 | ||
NM_117620.1 | NP_193267.1 | 5 - 6 | 1304 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |