Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036570.1 | NP_001031647.1 | 10 - 11 | 1539 | 86 | |
NM_001084926.1 | NP_001078395.1 | 9 - 10 | 1476 | 86 | ||
NM_001084927.2 | NP_001078396.1 | 8 - 9 | 1528 | 86 | ||
NM_001125515.1 | NP_001118987.1 | 8 - 9 | 1421 | 86 | ||
NM_001341051.1 | NP_001328869.1 | 8 - 9 | 1647 | 86 | ||
NM_117643.4 | NP_193288.2 | 7 - 8 | 1277 | 86 | ||
NM_179060.4 | NP_849391.2 | 9 - 10 | 1515 | 86 | ||
GenBank mRNA | AK317187.1 | - | 9 - 10 | 1504 | 86 | |
AK317578.1 | - | 7 - 8 | 1195 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |