Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341097.1 | NP_001319958.1 | 10 - 11 | 1704 | 69 | |
NM_117729.8 | NP_193367.7 | 9 - 10 | 1656 | 69 | ||
GenBank mRNA | AF465831.1 | - | 9 - 10 | 1589 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |