Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP70-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117827.3 | NP_193455.2 | 6 - 7 | 1036 | 81 | |
GenBank mRNA | AK117336.1 | - | 6 - 7 | 950 | 81 | |
AM086442.1 | - | 6 - 7 | 923 | 81 | ||
BT005901.1 | - | 6 - 7 | 865 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |