Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117857.3 | NP_193484.1 | 1 - 2 | 261 | 116 | |
GenBank mRNA | AK118904.1 | - | 1 - 2 | 200 | 116 | |
BT005488.1 | - | 1 - 2 | 78 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |