Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117980.5 | NP_193599.3 | 8 - 9 | 1818 | 74 | |
GenBank mRNA | FJ708745.1 | - | 7 - 8 | 1103 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |