Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117994.4 | NP_567564.1 | 11 - 12 | 2540 | 131 | |
GenBank mRNA | AY099636.1 | - | 11 - 12 | 2440 | 131 | |
KJ676982.1 | - | 11 - 12 | 2373 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |